Thermo Scientific Chemicals

4'-Isobutylacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: A10624.18
50 g, Each
Thermo Scientific Chemicals

4'-Isobutylacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: A10624.18
50 g, Each
Quantity
Catalog number: A10624.18
also known as A10624-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38861-78-8
IUPAC Name
1-[4-(2-methylpropyl)phenyl]ethan-1-one
Molecular Formula
C12H16O
InChI Key
KEAGRYYGYWZVPC-UHFFFAOYSA-N
SMILES
CC(C)CC1=CC=C(C=C1)C(C)=O
Form
Liquid
Refractive Index
1.5140-1.5190 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms

Description

4'-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.

Solubility
Miscible with chloroform and methanol. Slightly miscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with reducing agents, strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text