Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals
Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A10626.14
also known as A10626-14
Price (USD)/ Each
63.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
63.30
Add to cart
Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals
Catalog numberA10626.14
Price (USD)/ Each
63.30
-
Add to cart
Chemical Identifiers
CAS618-98-4
IUPAC Nameethyl 3-nitrobenzoate
Molecular FormulaC9H9NO4
InChI KeyMKBIJCPQTPFQKQ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)39.0-45.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only