Thermo Scientific Chemicals

Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A10626.22
100 g, Each
Thermo Scientific Chemicals

Ethyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A10626.22
100 g, Each
Quantity
Catalog number: A10626.22
also known as A10626-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-98-4
IUPAC Name
ethyl 3-nitrobenzoate
Molecular Formula
C9H9NO4
InChI Key
MKBIJCPQTPFQKQ-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Assay (GC)
≥98.0%
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
39.0-45.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text