Thermo Scientific Chemicals

Bis(cyclohexanone) oxaldihydrazone, 98%, Thermo Scientific Chemicals

Catalog number: A10628.14
25 g, Each
Thermo Scientific Chemicals

Bis(cyclohexanone) oxaldihydrazone, 98%, Thermo Scientific Chemicals

Catalog number: A10628.14
25 g, Each
Quantity
Catalog number: A10628.14
also known as A10628-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
370-81-0
IUPAC Name
N',N'-dicyclohexylideneethanedihydrazide
Molecular Formula
C14H22N4O2
InChI Key
DSRJIHMZAQEUJV-UHFFFAOYSA-N
SMILES
O=C(NN=C1CCCCC1)C(=O)NN=C1CCCCC1
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms
Form
Powder
Melting Point (clear melt)
208.0-214.0?C
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text