Thermo Scientific Chemicals

3-Chloropropanesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A10631.30
250 g, Each
Thermo Scientific Chemicals

3-Chloropropanesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A10631.30
250 g, Each
Quantity
Catalog number: A10631.30
also known as A10631-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1633-82-5
IUPAC Name
3-chloropropane-1-sulfonyl chloride
Molecular Formula
C3H6Cl2O2S
InChI Key
GPKDGVXBXQTHRY-UHFFFAOYSA-N
SMILES
ClCCCS(Cl)(=O)=O
Appearance (Color)
Clear colorless to yellow to orange or pale brown
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4880-1.4920 @ 20°C

Description

3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl Acetate and hexane.

Notes
Store at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Keep away from alcohols, strong bases, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text