Thermo Scientific Chemicals

6-Nitrobenzothiazole, 98+%, Thermo Scientific Chemicals

Catalog number: A10634.18
50 g, Each
Thermo Scientific Chemicals

6-Nitrobenzothiazole, 98+%, Thermo Scientific Chemicals

Catalog number: A10634.18
50 g, Each
Quantity
Catalog number: A10634.18
also known as A10634-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2942-06-5
IUPAC Name
6-nitro-1,3-benzothiazole
Molecular Formula
C7H4N2O2S
InChI Key
QLUFBCVWKTWKBF-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C2N=CSC2=C1
Assay (GC)
≥98.0%
Melting Point (clear melt)
171.5-177.5?C
Appearance (Color)
Yellow to pale brown to pale cream
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text