Thermo Scientific Chemicals

4-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10646.22
100 g, Each
Thermo Scientific Chemicals

4-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10646.22
100 g, Each
Quantity
Catalog number: A10646.22
also known as A10646-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
405-50-5
IUPAC Name
2-(4-fluorophenyl)acetic acid
Molecular Formula
C8H7FO2
InChI Key
MGKPFALCNDRSQD-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(F)C=C1
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to cream

Description

4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text