N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals
N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25g
5g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10647.14
also known as A10647-14
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
-
Add to cart
Quantity:
25g
Request bulk or custom format
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
Add to cart
N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals
Catalog numberA10647.14
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
-
Add to cart
Chemical Identifiers
CAS13139-15-6
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid
Molecular FormulaC11H21NO4
InChI KeyMDXGYYOJGPFFJL-SVGMAFHSNA-N
SMILESCC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥98.5% (U.S. specification)
Water Content (Karl Fischer Titration)7.0-9.0% (non-U.S. specification)
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (dry wt basis, non-U.S. specification)
FormCrystalline powder or powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only