4,4'-Difluorobenzophenone, 98+%, Thermo Scientific Chemicals
4,4'-Difluorobenzophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Difluorobenzophenone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10653.09
also known as A10653-09
Price (USD)/ Each
51.70
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
51.70
Add to cart
4,4'-Difluorobenzophenone, 98+%, Thermo Scientific Chemicals
Catalog numberA10653.09
Price (USD)/ Each
51.70
-
Add to cart
Chemical Identifiers
CAS345-92-6
IUPAC Namebis(4-fluorophenyl)methanone
Molecular FormulaC13H8F2O
InChI KeyLSQARZALBDFYQZ-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point105-111?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only