Thermo Scientific Chemicals

1-Bromopinacolone, 97+%, Thermo Scientific Chemicals

Catalog number: A10655.18
50 g, Each
Thermo Scientific Chemicals

1-Bromopinacolone, 97+%, Thermo Scientific Chemicals

Catalog number: A10655.18
50 g, Each
Quantity
Catalog number: A10655.18
also known as A10655-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5469-26-1
IUPAC Name
1-bromo-3,3-dimethylbutan-2-one
Molecular Formula
C6H11BrO
InChI Key
SAIRZMWXVJEBMO-UHFFFAOYSA-N
SMILES
CC(C)(C)C(=O)CBr
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4630-1.4680 @ 20?C
Form
Liquid
Assay (GC)
≥97.0%

Description

1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text