Thermo Scientific Chemicals

4-Bromobutyronitrile, 97%, Thermo Scientific Chemicals

Catalog number: A10674.30
250 g, Each
Thermo Scientific Chemicals

4-Bromobutyronitrile, 97%, Thermo Scientific Chemicals

Catalog number: A10674.30
250 g, Each
Quantity
Catalog number: A10674.30
also known as A10674-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5332-06-9
IUPAC Name
4-bromobutanenitrile
Molecular Formula
C4H6BrN
InChI Key
CQPGDDAKTTWVDD-UHFFFAOYSA-N
SMILES
BrCCCC#N
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Comment
Material sourced in the U.S. and in other countries
Refractive Index
1.4755-1.4805 @ 20°C (non-U.S. sourced material)

Description

4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.

Solubility
Miscible with toluene and benzene. Immiscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text