Thermo Scientific Chemicals

Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: A10677.14
25 g, Each
Thermo Scientific Chemicals

Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: A10677.14
25 g, Each
Quantity
Catalog number: A10677.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1075-06-5
IUPAC Name
2,2-dihydroxy-1-phenylethan-1-one
Molecular Formula
C8H8O3
InChI Key
NBIBDIKAOBCFJN-UHFFFAOYSA-N
SMILES
OC(O)C(=O)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown or pink
Assay (GC)
≥96.0%
Water Content (Karl Fischer Titration)
6.29-16.77% (0.5-1.5 waters)

Description

Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

Solubility
Partly miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text