Thermo Scientific Chemicals

D-Proline, 99%, Thermo Scientific Chemicals

Catalog number: A10682.03
1 g, Each
Thermo Scientific Chemicals

D-Proline, 99%, Thermo Scientific Chemicals

Catalog number: A10682.03
1 g, Each
Quantity
Catalog number: A10682.03
also known as A10682-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
344-25-2
IUPAC Name
pyrrolidine-2-carboxylic acid
Molecular Formula
C5H9NO2
InChI Key
ONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCN1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
85.6 ± 3.0° (C=1 in water)

Description

D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.

Solubility
Soluble in water (1620 g/L at 20°C), and acetone (slightly). Insoluble in ethanol.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text