Thermo Scientific Chemicals

DL-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A10683.36
500 g, Each
Thermo Scientific Chemicals

DL-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A10683.36
500 g, Each
Quantity
Catalog number: A10683.36
also known as A10683-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
133-37-9
IUPAC Name
2,3-dihydroxybutanedioic acid
Molecular Formula
C4H6O6
InChI Key
FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES
OC(C(O)C(O)=O)C(O)=O
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Silylated GC)
≥98.5%

Description

DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.

Solubility
Soluble in water, alcohol, mineral acids and alkalies.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text