Thermo Scientific Chemicals

Phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A10694.22
100 g, Each
Thermo Scientific Chemicals

Phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A10694.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10694.22
also known as A10694-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-29-4
Form
Liquid
Identification (FTIR)
Conforms (UK sourced material)
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0% (UK sourced material)
Comment
Material sourced in UK and US

Description

Phenylacetonitrile is used in organic synthesis for dyes, perfumes, pharmaceuticals, especially penicillin precursors. It is also used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text