Thermo Scientific Chemicals

1-Bromobutane, 98+%, Thermo Scientific Chemicals

Catalog number: A10696.0C
10,000 g, Each
Thermo Scientific Chemicals

1-Bromobutane, 98+%, Thermo Scientific Chemicals

Catalog number: A10696.0C
10,000 g, Each
Quantity
Catalog number: A10696.0C
also known as A10696-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-65-9
IUPAC Name
1-bromobutane
Molecular Formula
C4H9Br
InChI Key
MPPPKRYCTPRNTB-UHFFFAOYSA-N
SMILES
CCCCBr
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4380-1.4420 @ 20?C
Form
Liquid

Description

1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text