2-Adamantanol, 98%, Thermo Scientific Chemicals
2-Adamantanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Adamantanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A10697.14
also known as A10697-14
Price (USD)/ Each
189.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
189.00
Add to cart
2-Adamantanol, 98%, Thermo Scientific Chemicals
Catalog numberA10697.14
Price (USD)/ Each
189.00
-
Add to cart
Chemical Identifiers
CAS700-57-2
IUPAC Nameadamantan-2-ol
Molecular FormulaC10H16O
InChI KeyFOWDOWQYRZXQDP-UHFFFAOYSA-N
SMILESOC1C2CC3CC(C2)CC1C3
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only