Thermo Scientific Chemicals

(1R)-(+)-Camphor, 98%, Thermo Scientific Chemicals

Catalog number: A10708.14
25 g, Each
Thermo Scientific Chemicals

(1R)-(+)-Camphor, 98%, Thermo Scientific Chemicals

Catalog number: A10708.14
25 g, Each
Quantity
Catalog number: A10708.14
also known as A10708-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
464-49-3
IUPAC Name
(1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular Formula
C10H16O
InChI Key
DSSYKIVIOFKYAU-XVKPBYJWSA-N
SMILES
CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2
Appearance (Color)
White
Melting Point (clear melt)
174.0-180.0°C
Optical Rotation
+41.0 to +45.5° (c=10 in ethanol)
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps or fused solid

Description

(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text