Thermo Scientific Chemicals

Ethyl potassium malonate, 98%, Thermo Scientific Chemicals

Catalog number: A10720.22
100 g, Each
Thermo Scientific Chemicals

Ethyl potassium malonate, 98%, Thermo Scientific Chemicals

Catalog number: A10720.22
100 g, Each
Quantity
Catalog number: A10720.22
also known as A10720-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6148-64-7
IUPAC Name
3-ethoxy-3-oxopropanoic acid potassium
Molecular Formula
C5H8KO4
InChI Key
HRBCMCJOBRIIQG-UHFFFAOYSA-N
SMILES
[K].CCOC(=O)CC(O)=O
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Ethyl potassium malonate is used as a competitive inhibitor of the enzyme succinate dehydrogenase. It acts as a precursor to produce (trimethylsilyl)ethyl malonate, which is utilized to prepare beta-ketoesters by acylation. Further, it reacts with aryl nitriles to prepare beta-amino acrylates in the presence of zinc chloride and a catalytic amount of Hünig's base. In addition to this, it serves as an intermediate for the preparation of ethyl tert-butyl malonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl potassium malonate is used as a competitive inhibitor of the enzyme succinate dehydrogenase. It acts as a precursor to produce (trimethylsilyl)ethyl malonate, which is utilized to prepare beta-ketoesters by acylation. Further, it reacts with aryl nitriles to prepare beta-amino acrylates in the presence of zinc chloride and a catalytic amount of Hünig′s base. In addition to this, it serves as an intermediate for the preparation of ethyl tert-butyl malonate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text