3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10723.06
also known as A10723-06
Price (USD)/ Each
32.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
32.50
Add to cart
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Catalog numberA10723.06
Price (USD)/ Each
32.50
-
Add to cart
Chemical Identifiers
CAS6099-04-3
IUPAC Name(2E)-3-(3-methoxyphenyl)prop-2-enoic acid
Molecular FormulaC10H10O3
InChI KeyLZPNXAULYJPXEH-AATRIKPKSA-N
SMILESCOC1=CC=CC(C=CC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to cream to pale orange
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)115.0-121.0°C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only