Thermo Scientific Chemicals

4-Iodobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10730.09
10 g, Each
Thermo Scientific Chemicals

4-Iodobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10730.09
10 g, Each
Quantity
Catalog number: A10730.09
also known as A10730-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
619-58-9
IUPAC Name
4-iodobenzoic acid
Molecular Formula
C7H5IO2
InChI Key
GHICCUXQJBDNRN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(I)C=C1
Assay (Silylated GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
267.0-276.0?C
Appearance (Color)
White to pale cream to pale orange to pale brown
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Partly miscible with water. Soluble in alcohol & ether.

Notes
It is sensitive to light. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text