Thermo Scientific Chemicals

Benzophenone, 99%, Thermo Scientific Chemicals

Catalog number: A10739.0I
5000 g, Each
Thermo Scientific Chemicals

Benzophenone, 99%, Thermo Scientific Chemicals

Catalog number: A10739.0I
5000 g, Each
Quantity
Catalog number: A10739.0I
also known as A10739-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-61-9
IUPAC Name
diphenylmethanone
Molecular Formula
C13H10O
InChI Key
RWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILES
O=C(C1=CC=CC=C1)C1=CC=CC=C1
Assay (GC)
≥98.5% (UK Source Material)
Comment
Material Sourced in UK and US
Form
Crystalline powder or flakes
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥98.5% (US Sourced Material)

Description

Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.

Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and reducing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text