Thermo Scientific Chemicals

1-Cyclohexene-1-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10741.06
5 g, Each
Thermo Scientific Chemicals

1-Cyclohexene-1-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10741.06
5 g, Each
Quantity
Catalog number: A10741.06
also known as A10741-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
636-82-8
IUPAC Name
cyclohex-1-ene-1-carboxylic acid
Molecular Formula
C7H10O2
InChI Key
NMEZJSDUZQOPFE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CCCCC1
Appearance (Color)
Colorless or white to pale cream or pale yellow
Form
Powder or crystalline powder or fused solid or clear liquid as melt
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms

Description

1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

Solubility
Soluble in water (0.7 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text