Thermo Scientific Chemicals

Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A10744.14
25 g, Each
Thermo Scientific Chemicals

Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A10744.14
25 g, Each
Quantity
Catalog number: A10744.14
also known as A10744-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
336-59-4
IUPAC Name
2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular Formula
C8F14O3
InChI Key
UFFSXJKVKBQEHC-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Identification (FTIR)
Conforms
Refractive Index
<1.3000 @ 20°C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.

Solubility
Miscible with halogenated solvents. Immiscible with polar solvents.

Notes
Incompatible with moist air, water, strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text