2-Bromoisovaleric acid, 96%, Thermo Scientific Chemicals
2-Bromoisovaleric acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoisovaleric acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A10756.18
also known as A10756-18
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
2-Bromoisovaleric acid, 96%, Thermo Scientific Chemicals
Catalog numberA10756.18
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS565-74-2
IUPAC Name2-bromo-3-methylbutanoic acid
Molecular FormulaC5H9BrO2
InChI KeyUEBARDWJXBGYEJ-UHFFFAOYNA-N
SMILESCC(C)C(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Melting Point (clear melt)33.0-42.0?C
FormFused solid
Assay (Silylated GC)≥95.0%
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only