Thermo Scientific Chemicals

3-Methoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10767.18
50 g, Each
Thermo Scientific Chemicals

3-Methoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10767.18
50 g, Each
Quantity
Catalog number: A10767.18
also known as A10767-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-38-9
IUPAC Name
3-methoxybenzoic acid
Molecular Formula
C8H8O3
InChI Key
XHQZJYCNDZAGLW-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C(O)=O
Appearance (Color)
White to cream
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
103.0-109.0?C
Form
Crystals or powder or crystalline powder or lumps
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

Solubility
Soluble in 95% ethanol (50 mg/ml), water (2 mg/ml at 25°C) and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text