3-Methoxybenzoic acid, 99%, Thermo Scientific Chemicals
3-Methoxybenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxybenzoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A10767.30
also known as A10767-30
Price (USD)/ Each
247.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
247.00
Add to cart
3-Methoxybenzoic acid, 99%, Thermo Scientific Chemicals
Catalog numberA10767.30
Price (USD)/ Each
247.00
-
Add to cart
Chemical Identifiers
CAS586-38-9
IUPAC Name3-methoxybenzoic acid
Molecular FormulaC8H8O3
InChI KeyXHQZJYCNDZAGLW-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)≥98.5%
Melting Point (clear melt)103.0-109.0?C
FormCrystals or powder or crystalline powder or lumps
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

Solubility
Soluble in 95% ethanol (50 mg/ml), water (2 mg/ml at 25°C) and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Nicolay Dodoff.; Konstantin Grancharov.; Rumyana Gugova.; Nadejda Spassovska. Platinum (II) complexes of benzoic- and 3-methoxybenzoic acid hydrazides. Synthesis, characterization, and cytotoxic effect. Journal of Inorganic Biochemistry. 1994, 54, (3), 221-233.
  2. K A DeWeerd.; A Saxena, D P Nagle Jr.; J M Suflita. Metabolism of the 18O-methoxy substituent of 3-methoxybenzoic acid and other unlabeled methoxybenzoic acids by anaerobic bacteria. Appl. Environ. Microbiol. 1988, 54 1237-1242.