Triethylene glycol, 99%, Thermo Scientific Chemicals
Triethylene glycol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylene glycol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A10772.0D
also known as A10772-0D
Price (USD)/ Each
227.65
Online exclusive
253.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
10,000 mL
Request bulk or custom format
Price (USD)/ Each
227.65
Online exclusive
253.00 
Save 25.35 (10%)
Add to cart
Triethylene glycol, 99%, Thermo Scientific Chemicals
Catalog numberA10772.0D
Price (USD)/ Each
227.65
Online exclusive
253.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS112-27-6
IUPAC Name2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
Molecular FormulaC6H14O4
InChI KeyZIBGPFATKBEMQZ-UHFFFAOYSA-N
SMILESOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial sourced in UK and US
Refractive Index1.4540-1.4575 @ 20°C (UK-sourced material)
Water Content (Karl Fischer Titration)≤0.5%
FormLiquid
Assay from Supplier's CofA≥98.5% (US-sourced material)
View more
Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

Solubility
Miscible with water, ethanol, acetone, benzene, toluene, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil, fat, petroleum ether and most hydrocarbons.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
RUO – Research Use Only

General References:

  1. Gutiérrez-Alejandre, A.; Laurrabaquio-Rosas, G.; Ramírez, J.; Busca, G. On the role of triethylene glycol in the preparation of highly active Ni-Mo/Al2O3 hydrodesulfurization catalysts: A spectroscopic study. Appl. Catal., B 2015, 166-167, 560-567.
  2. Wang, Q.; Song, W. L.; Fan, L. Z.; Shi, Q. Effect of alumina on triethylene glycol diacetate-2-propenoic acid butyl ester composite polymer electrolytes for flexible lithium ion batteries. J. Power Sources 2015, 279, 405-412.