Thermo Scientific Chemicals

Trimethylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10776.0E
2500 g, Each
Thermo Scientific Chemicals

Trimethylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10776.0E
2500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10776.0E
also known as A10776-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-98-9
IUPAC Name
2,2-dimethylpropanoic acid
Molecular Formula
C5H10O2
InChI Key
IUGYQRQAERSCNH-UHFFFAOYSA-N
SMILES
CC(C)(C)C(O)=O
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder or fused solid
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream

Description

Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

Solubility
Soluble in alcohol, ether. Slightly soluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text