Trimethylacetic acid, 99%, Thermo Scientific Chemicals
Trimethylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
10,000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10776.36
also known as A10776-36
Price (USD)/ Each
55.65
Online exclusive
62.00 
Save 6.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
55.65
Online exclusive
62.00 
Save 6.35 (10%)
Add to cart
Trimethylacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberA10776.36
Price (USD)/ Each
55.65
Online exclusive
62.00 
Save 6.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-98-9
IUPAC Name2,2-dimethylpropanoic acid
Molecular FormulaC5H10O2
InChI KeyIUGYQRQAERSCNH-UHFFFAOYSA-N
SMILESCC(C)(C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic acid can be used as an internal chemical-shift standard for NMR of aqueous solutions. It is a metabolite of oral cephem (β-lactam) antibiotics such as S-1108, containing pivaloyl moieties.

Solubility
Soluble in alcohol, ether. Slightly soluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. S.Albert; H.S.Gutowsky; J.A.Ripmeester. NMR investigations of solid (CH3)3CCOOH (trimethylacetic acid) and (CH3)3CCOOD. J. Chem. Phys. 1976, 64, (8),3277
  2. Alfred L.Miller; Newell C.Cook; Frank C.Whitmore. The Ketonic Decarboxylation Reaction1: The Ketonic Decarboxylation of Trimethylacetic Acid2 and Isobutyric Acid. J. Am. Chem. Soc. 1950, 72, (6),2732-2735
  3. In the presence of an Ag(II) species (from silver nitrate and ammonium persulfate in TFA), tert-butyl radicals are generated. With quinoline, the 2-tert-butyl substituted derivative is formed in high yield: Tetrahedron, 46, 2525 (1990), whereas for most other carboxylic acids, both 2- and 4-substitution usually occur.