Thermo Scientific Chemicals

7,7,8,8-Tetracyanoquinodimethane, 98%, Thermo Scientific Chemicals

Catalog number: A10779.14
25 g, Each
Thermo Scientific Chemicals

7,7,8,8-Tetracyanoquinodimethane, 98%, Thermo Scientific Chemicals

Catalog number: A10779.14
25 g, Each
Quantity
Catalog number: A10779.14
also known as A10779-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1518-16-7
IUPAC Name
2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular Formula
C12H4N4
InChI Key
PCCVSPMFGIFTHU-UHFFFAOYSA-N
SMILES
N#CC(C#N)=C1C=CC(C=C1)=C(C#N)C#N
Appearance (Color)
Yellow to dark green to green to orange to brown
Form
Crystals or powder or crystalline powder or fused solid
Assay (HPLC)
≥97.5%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)

Description

7,7,8,8-Tetracyanoquinodimethane is an electron-acceptor molecule used to form charge-transfer superconductors. It is an effective catalyst used for the ?-chlorination of carboxylic acids using chlorosulfonic acid; the presence of TCNQ suppresses competing free-radical chlorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7,7,8,8-Tetracyanoquinodimethane is an electron-acceptor molecule used to form charge-transfer superconductors. It is an effective catalyst used for the ɑ-chlorination of carboxylic acids using chlorosulfonic acid; the presence of TCNQ suppresses competing free-radical chlorination.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text