Thermo Scientific Chemicals

Adenosine, 99%

Catalog number: A10781.30
250 g, Each
Thermo Scientific Chemicals

Adenosine, 99%

Catalog number: A10781.30
250 g, Each
Quantity
Catalog number: A10781.30
also known as A10781-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-61-7
IUPAC Name
(2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Formula
C10H13N5O4
InChI Key
OIRDTQYFTABQOQ-KQYNXXCUSA-N
SMILES
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
-70.0 ± 2.0? (C=2 in 5% NaOH)
Form
Powder
Identification (FTIR)
Conforms

Description

Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text