Thermo Scientific Chemicals

3-Methylbenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10784.22
100 g, Each
Thermo Scientific Chemicals

3-Methylbenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10784.22
100 g, Each
Quantity
Catalog number: A10784.22
also known as A10784-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-19-9
IUPAC Name
1-(chloromethyl)-3-methylbenzene
Molecular Formula
C8H9Cl
InChI Key
LZBOHNCMCCSTJX-UHFFFAOYSA-N
SMILES
CC1=CC=CC(CCl)=C1
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5330-1.5370 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text