Informational:NEW - Major Software Update Available! — Cell Cycle Analysis can be performed using the Tali® Image-Based Cytometer
Invitrogen™

Tali™ Viability Kit - Dead Cell Red

Catalog number: A10786
Have Questions?
Invitrogen™

Tali™ Viability Kit - Dead Cell Red

Catalog number: A10786
Have Questions?
Catalog Number
A10786
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A107861 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Tali™ Viability Kit - Dead Cell Red is a ready-to-use solution of propidium iodide (PI) that has been validated for use with the Tali™ Image-Based Cytometer. PI is a live cell-impermeant fluorogenic DNA-binding dye that has been extensively used to identify necrotic cells.

Key Features of Tali™ Viability Kit - Dead Cell Red:

• Impermeant to live cells
• Easily enters and binds to nucleic acids in necrotic cells

After staining a cell population using Dead Cell Red™, dead cells display red fluorescence upon excitation. Tali™ Viability Kit - Dead Cell Red can be used to identify dead cells in a population of unstained cells, or in a population of GFP-expressing cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Cell Viability Kit
Cell Type
Eukaryotic Cell
Detection Method
Fluorescent
For Use With (Equipment)
Tali™ Image-Based Cytometer
Compatible Cells
Eukaryotic Cells
Label or Dye
Propidium Iodide
Shipping Condition
Room Temperature
Format
Vial
Product Line
Tali™

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text