Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%, Thermo Scientific Chemicals
Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A10790.14
also known as A10790-14
Price (USD)/ Each
654.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
654.00
Add to cart
Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%, Thermo Scientific Chemicals
Catalog numberA10790.14
Price (USD)/ Each
654.00
-
Add to cart
Chemical Identifiers
CAS43088-42-2
IUPAC Nameethyl 2-amino-4-methylthiophene-3-carboxylate
Molecular FormulaC8H11NO2S
InChI KeyILYCZKOBLRJJSW-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(N)SC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)78.0-84.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only