Thermo Scientific™

Hydrazine sulfate, 99+%, Thermo Scientific Chemicals

Catalog number: A10792.A1
1 kg, Each
Thermo Scientific™

Hydrazine sulfate, 99+%, Thermo Scientific Chemicals

Catalog number: A10792.A1
1 kg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10792.A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-93-2
IUPAC Name
hydrazine; sulfuric acid
Molecular Formula
H6N2O4S
InChI Key
ZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILES
NN.OS(O)(=O)=O
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (Redox Titration)
≥99.0% to ≤101.0%

Description

Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrazine sulfate is used in analytical chemistry for the gravimetric estimation of nickel, cobalt and cadmium. It is also used in the refining of rare metals as well as in the separation of polonium from tellurium. It finds application in the synthesis of chemical intermediate and organic compounds. Further, it acts as an antioxidant in soldering flux for light metals and as a precursor to hydrazine.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents and bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text