Thermo Scientific Chemicals

Methyl 3,5-dibromo-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10799.18
50 g, Each
Thermo Scientific Chemicals

Methyl 3,5-dibromo-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10799.18
50 g, Each
Quantity
Catalog number: A10799.18
also known as A10799-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41727-47-3
IUPAC Name
methyl 3,5-dibromo-4-hydroxybenzoate
Molecular Formula
C8H6Br2O3
InChI Key
NVGJGYZKXBLIKY-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Br)=C(O)C(Br)=C1
Appearance (Color)
White
Form
Powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
119.0-125.0?C

Description

Methyl 3,5-dibromo-4-hydroxybenzoate is used to produce 4-hydroxy-3,5-di-pyridin-2-yl-benzoic acid methyl ester at temperature of 90-100°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-dibromo-4-hydroxybenzoate is used to produce 4-hydroxy-3,5-di-pyridin-2-yl-benzoic acid methyl ester at temperature of 90-100°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text