Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A10807.03
1 g, Each
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A10807.03
1 g, Each
Quantity
Catalog number: A10807.03
also known as A10807-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25952-53-8
IUPAC Name
({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Molecular Formula
C8H18ClN3
InChI Key
FPQQSJJWHUJYPU-UHFFFAOYSA-N
SMILES
Cl.CCN=C=NCCCN(C)C
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥98.0%
Appearance (Color)
White
Assay (Titration ex Chloride)
≥98.0 to ≤102.0% (non-U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries

Description

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3'-amino-3'-deoxyadenosine-5'-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in acetonitrile dichloromethane, dimethylformamide, and tetrahydrofuran. Slightly soluble in water.

Notes
Sensitive to moisture. Incompatible with strong acids, strong oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text