1-(2-Thenoyl)-3,3,3-trifluoroacetone is widely used in the analysis of metals such as actinides and lanthanides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Thenoyl)-3,3,3-trifluoroacetone is widely used in the analysis of metals such as actinides and lanthanides.
Solubility
Soluble in methanol and ethanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Chelating ligand. Reagent for Ce, Co, Cr, Cu, Fe, U, V.
- Kedari, C. S.; Pandita, S. S.; Parikha, K. J.; Tripathi, S. C.; Gandhi, P. M. Extraction of plutonium (IV) from aqueous nitrate solutions into ligand modified micellar phase (LMMP) of Tergitol 15-S-9 with tri-octylphosphine oxide and separation by ultrafiltration. Desalin. Water Treat. 2014, 52 (1-3), 446-451.
- Oliveira, T. C.; Monteiro, R. P. G.; Kastner, G. F.; Barbier, F. B.; Oliveira, A. H. Radiochemical methodologies applied to determination of zirconium isotopes in low-level waste samples from nuclear power plants. J. Radioanal. Nucl. Chem. 2014, 302 (1), 41-47.