Thermo Scientific Chemicals

N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals

Catalog number: A10810.06
5g, Each
Thermo Scientific Chemicals

N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals

Catalog number: A10810.06
5g, Each
Quantity
Catalog number: A10810.06
also known as A10810-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3978-80-1
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C14H19NO5
InChI Key
CNBUSIJNWNXLQQ-LDGXTIHJNA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Comment
Specification differs for U.S. and non-U.S. material where indicated
Optical Rotation
34.0-38.0° (C=1, Dioxane, U.S. specification)
Optical Rotation
37.0 ± 0.1° (C=1 in dioxane, non-U.S. specification)
Appearance (Color)
White to pale cream
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text