Thermo Scientific Chemicals

4-Phenylphenol, 97%, Thermo Scientific Chemicals

Catalog number: A10817.0E
2500 g, Each
Thermo Scientific Chemicals

4-Phenylphenol, 97%, Thermo Scientific Chemicals

Catalog number: A10817.0E
2500 g, Each
Quantity
Catalog number: A10817.0E
also known as A10817-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-69-3
IUPAC Name
[1,1'-biphenyl]-4-ol
Molecular Formula
C12H10O
InChI Key
YXVFYQXJAXKLAK-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream to pale brown or pale pink
Melting Point (clear melt)
163.0-170.0°C
Assay (GC)
≥96.0%
Form
Powder and/or flakes

Description

Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text