Thermo Scientific Chemicals

4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A10823.14
25 g, Each
Thermo Scientific Chemicals

4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A10823.14
25 g, Each
Quantity
Catalog number: A10823.14
also known as A10823-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-83-0
IUPAC Name
4-chlorobenzene-1,2-diamine
Molecular Formula
C6H7ClN2
InChI Key
BXIXXXYDDJVHDL-UHFFFAOYSA-N
SMILES
NC1=CC=C(Cl)C=C1N
Form
Powder
Melting Point (clear melt)
68.0-77.0?C
Appearance (Color)
Pale brown to brown
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text