Thermo Scientific Chemicals

Bromotrichloromethane, 97%, Thermo Scientific Chemicals

Catalog number: A10824.36
500 g, Each
Thermo Scientific Chemicals

Bromotrichloromethane, 97%, Thermo Scientific Chemicals

Catalog number: A10824.36
500 g, Each
Quantity
Catalog number: A10824.36
also known as A10824-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-62-7
IUPAC Name
bromotrichloromethane
Molecular Formula
CBrCl3
InChI Key
XNNQFQFUQLJSQT-UHFFFAOYSA-N
SMILES
ClC(Cl)(Cl)Br
Identification (FTIR)
Conforms
Refractive Index
1.5040-1.5090 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%

Description

Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Protect against electrostatic charges.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text