Thermo Scientific Chemicals

Hydroxyurea, 98%, Thermo Scientific Chemicals

Catalog number: A10831.03
1 g, Each
Thermo Scientific Chemicals

Hydroxyurea, 98%, Thermo Scientific Chemicals

Catalog number: A10831.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10831.03
also known as A10831-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-07-1
IUPAC Name
hydroxyurea
Molecular Formula
CH4N2O2
InChI Key
VSNHCAURESNICA-UHFFFAOYSA-N
SMILES
NC(=O)NO
Appearance (Color)
White to cream or yellow
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Proton NMR
Conforms to structure (non-U.S. specification)
Form
Crystals or powder or crystalline powder

Description

Hydroxyurea is used as an inhibitor of DNA synthesis and an anti-cancer drug. It is used as a precursor to prepare 1-hydroxy-5,6-dimethyl-1H-pyrimidine-2,4-dione via reaction with 2-methyl-3-oxo-butyric acid ethyl ester. It is used in the treatment of melanoma, resistant chronic myelocytic leukemia and recurrent metastatic or inoperable ovarian. It is also involved in reducing the rate of painful attacks in sickle-cell disease and has anti-retroviral properties in diseases such as AIDS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, hot alcohol and dimethyl sulfoxide. Insoluble in ethanol and benzene.

Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text