N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10833.22
also known as A10833-22
Price (USD)/ Each
14.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
17.40 
Save 2.75 (16%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
14.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
17.40 
Save 2.75 (16%)
Add to cart
N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
Catalog numberA10833.22
Price (USD)/ Each
14.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
17.40 
Save 2.75 (16%)
-
Add to cart
Chemical Identifiers
CAS111-41-1
IUPAC Name2-[(2-aminoethyl)amino]ethan-1-ol
Molecular FormulaC4H12N2O
InChI KeyLHIJANUOQQMGNT-UHFFFAOYSA-N
SMILESNCCNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
Assay from Suppliers CofA≥98.5%
FormLiquid
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

Solubility
Fully miscible in water.

Notes
Air Sensitive & Hygroscopic. Store under inert gas. Keep away from incompatibles.
RUO – Research Use Only

General References:

  1. Hassan Ali Zamani.; Mohammad Taghi Hamed-Mosavian.;Elham Hamidfar.; Mohammad Reza Ganjali.; Parviz Norouzi. A novel iron (III)-PVC membrane potentiomeric sensor based on N-(2-hydroxyethyl)ethylenediamine-N,N',N″-triacetic acid. Materials Science and Engineering: C. 2008, 28, (8), 1551-1555.
  2. Hiroshi Ogino.; Mamoru Shimoi.; Yoko Saito. Structural identification of the reactive vanadium(III) intermediate formed in the electron-transfer reactions of [N'-(2-hydroxyethyl)ethylenediamine-N,N,N'-triacetato]aquavanadium(III) complex ([V(hedtra)(H2O)]) with halopentaamminecobalt(III) complexes: x-ray crystal structures of [V(hedtra)(H2O)].cntdot.2H2O and K[VO(hedtra)].cntdot.H2O. norg. Chem 1989, 28, (18), 3596-3600.