Thermo Scientific Chemicals

1-Methylpiperazine, 98+%, Thermo Scientific Chemicals

Catalog number: A10837.AP
500 mL, Each
Thermo Scientific Chemicals

1-Methylpiperazine, 98+%, Thermo Scientific Chemicals

Catalog number: A10837.AP
500 mL, Each
Quantity
Catalog number: A10837.AP
also known as A10837-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-01-3
IUPAC Name
1-methylpiperazine
Molecular Formula
C5H12N2
InChI Key
PVOAHINGSUIXLS-UHFFFAOYSA-N
SMILES
CN1CCNCC1
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4635-1.4675 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

1-Methylpiperazine is used in the preparation of 2-(4-methyl-1-piperazinylmethyl)acrylophenone. It is involved in the preparation of 1-(4-methoxy-phenyl)-4-methyl-piperazine by reacting with 1-chloro-4-methoxy-benzene. It acts as an intermediate in the synthesis of active pharmaceutical ingredients like ofloxacin, rifampicin, clozapine, sildenafil, trifluoperazine and zopiclone. Further, it is used as a mimic template in the preparation of molecularly imprinted microspheres. In addition to this, it is used in the generation of difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpiperazine is used in the preparation of 2-(4-methyl-1-piperazinylmethyl)acrylophenone. It is involved in the preparation of 1-(4-methoxy-phenyl)-4-methyl-piperazine by reacting with 1-chloro-4-methoxy-benzene. It acts as an intermediate in the synthesis of active pharmaceutical ingredients like ofloxacin, rifampicin, clozapine, sildenafil, trifluoperazine and zopiclone. Further, it is used as a mimic template in the preparation of molecularly imprinted microspheres. In addition to this, it is used in the generation of difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

Solubility
Miscible with water and methanol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text