Thermo Scientific Chemicals

3-Acetylbenzonitrile, 97+%, Thermo Scientific Chemicals

Catalog number: A10841.14
25 g, Each
Thermo Scientific Chemicals

3-Acetylbenzonitrile, 97+%, Thermo Scientific Chemicals

Catalog number: A10841.14
25 g, Each
Quantity
Catalog number: A10841.14
also known as A10841-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6136-68-1
Appearance (Color)
White to cream or pale yellow to pale brown
Form
Powder
Assay (GC)
≥97.0%
Melting Point (clear melt)
94.0-103.0°C

Description

Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text