Thermo Scientific Chemicals

Dibenzyl malonate, 95%, Thermo Scientific Chemicals

Catalog number: A10844.09
10 g, Each
Thermo Scientific Chemicals

Dibenzyl malonate, 95%, Thermo Scientific Chemicals

Catalog number: A10844.09
10 g, Each
Quantity
Catalog number: A10844.09
also known as A10844-09
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
15014-25-2
IUPAC Name
1,3-dibenzyl propanedioate
Molecular Formula
C17H16O4
InChI Key
RYFCSKVXWRJEOB-UHFFFAOYSA-N
SMILES
O=C(CC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Refractive Index
1.5395-1.5455 @ 20?C
Assay (GC)
≥94.0%

Description

Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl­ malonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl­ malonate.

Solubility
Sparingly soluble in water; miscible in all proportions with ether and alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text