3-Amino-1-phenyl-2-pyrazolin-5-one, 97%, Thermo Scientific Chemicals
3-Amino-1-phenyl-2-pyrazolin-5-one, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-1-phenyl-2-pyrazolin-5-one, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A10862.22
also known as A10862-22
Price (USD)/ Each
87.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
87.70
Add to cart
3-Amino-1-phenyl-2-pyrazolin-5-one, 97%, Thermo Scientific Chemicals
Catalog numberA10862.22
Price (USD)/ Each
87.70
-
Add to cart
Chemical Identifiers
CAS4149-06-8
IUPAC Name3-amino-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Molecular FormulaC9H9N3O
InChI KeyLPOVZHYARSAVIZ-UHFFFAOYSA-N
SMILESNC1=NN(C(=O)C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Cream to brown
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only