Thermo Scientific Chemicals

1,8-Diiodooctane, 97+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A10867.14
25 g, Each
Thermo Scientific Chemicals

1,8-Diiodooctane, 97+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A10867.14
25 g, Each
Quantity
Catalog number: A10867.14
also known as A10867-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24772-63-2
IUPAC Name
1,8-diiodooctane
Molecular Formula
C8H16I2
InChI Key
KZDTZHQLABJVLE-UHFFFAOYSA-N
SMILES
ICCCCCCCCI
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms
Stabilizer
Copper powder
Assay (GC)
≥97.0%
Refractive Index
1.5610-1.5660 @ 20?C

Description

1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

Solubility
Miscible with methanol.

Notes
Light sensitive. Incompatible with water and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text